Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013139.1 | NP_001013157.1 | 9 | 1601 | ||
XM_006253825.2 | XP_006253887.1 | 8 | 1659 | |||
GenBank mRNA | BC078956.1 | - | 9 | 1601 | 7110464 | |
FQ232145.1 | - | 8 | 1578 |