Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013742.3 | NP_001013764.1 | 26 | 3622 | ||
XM_017029268.1 | XP_016884757.1 | 25 | 3647 | |||
GenBank mRNA | AB183864.1 | - | 26 | 3622 | ||
BC137319.1 | - | 26 | 3606 | 9021316 | ||
BC137320.1 | - | 26 | 3606 | 9021317 | ||
DB728104.1 | - | 4 | 396 |