Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025127.1 | NP_001020298.1 | 6 | 1043 | ||
XM_006250781.2 | XP_006250843.1 | 6 | 1907 | |||
XM_006250782.2 | XP_006250844.1 | 6 | 2116 | |||
XM_006250783.2 | XP_006250845.1 | 6 | 1906 | |||
XM_006250784.2 | XP_006250846.1 | 5 | 632 | |||
XM_006250785.2 | XP_006250847.1 | 5 | 826 | |||
XM_008770044.1 | XP_008768266.1 | 5 | 844 | |||
XM_008770045.1 | XP_008768267.1 | 5 | 844 | |||
XM_008770046.1 | XP_008768268.1 | 6 | 2101 | |||
XM_008770047.1 | XP_008768269.1 | 5 | 844 | |||
XM_008770048.1 | XP_008768270.1 | 7 | 1960 | |||
GenBank mRNA | BC089952.1 | - | 6 | 1043 | 7312240 | |
EF538802.1 | - | 6 | 648 | |||
EF577045.1 | - | 5 | 594 |