Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021363.2 | NP_067338.2 | 3 | 1119 | ||
GenBank mRNA | AF242220.1 | - | 3 | 1104 | ||
AK020575.1 | - | 3 | 1116 | |||
DV048837.1 | - | 2 | 687 |