Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290765.1 | NP_001277694.1 | 10 | 1928 | ||
NM_026721.3 | NP_080997.1 | 10 | 1889 | |||
GenBank mRNA | AK003191.1 | - | 6 | 1192 | ||
AK035296.1 | - | 10 | 1796 | |||
AK211214.1 | - | 1 | 19 | |||
BC020106.1 | - | 8 | 1411 |