Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053805.1 | NP_446257.1 | 3 | 1453 | ||
GenBank mRNA | BC079210.1 | - | 3 | 1453 | 7131806 |