Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146775.1 | NP_666986.1 | 1 | 662 | ||
GenBank mRNA | BC148218.1 | - | 1 | 666 | 40134717 |