Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018128.4 | NP_060598.3 | 5 | 1803 | ||
GenBank mRNA | AB037822.1 | - | 5 | 1926 | ||
AK001396.1 | - | 3 | 445 | |||
AK026565.1 | - | 5 | 876 | |||
BC019090.2 | - | 4 | 711 | |||
BC110851.1 | - | 5 | 1381 | 5786818 | ||
BC126110.1 | - | 5 | 1351 | 8991826 |