Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015551.1 | NP_056366.1 | 5 | 3676 | ||
XM_005265034.3 | XP_005265091.1 | 4 | 3224 | |||
XM_017006137.1 | XP_016861626.1 | 5 | 3626 | |||
GenBank mRNA | AB011099.1 | - | 5 | 3676 |