Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018229.3 | NP_060699.3 | 2 | 1048 | ||
XM_006720196.2 | XP_006720259.1 | 2 | 677 | |||
XM_011536940.2 | XP_011535242.1 | 3 | 1020 | |||
GenBank mRNA | AF094583.1 | - | 1 | 13 | ||
AK001675.1 | - | 2 | 893 | |||
AK172824.1 | - | 1 | 110 | |||
AK225345.1 | - | 2 | 895 | |||
AK303999.1 | - | 3 | 922 | |||
AL136685.1 | - | 2 | 1027 | |||
BC013174.1 | - | 2 | 1012 | 3924075 |