Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 12 | 1655 | ||
NM_001143997.1 | NP_001137469.1 | 11 | 1450 | |||
NM_018245.2 | NP_060715.2 | 13 | 1826 | |||
XM_011539946.1 | XP_011538248.1 | 13 | 1711 | |||
XM_011539947.1 | XP_011538249.1 | 13 | 1917 | |||
XM_017016401.1 | XP_016871890.1 | 13 | 2101 | |||
XM_017016402.1 | XP_016871891.1 | 13 | 1728 | |||
XM_017016403.1 | XP_016871892.1 | 11 | 1474 | |||
GenBank mRNA | AB033116.3 | - | 12 | 1689 | ||
AK001713.1 | - | 13 | 1770 | |||
AK290226.1 | - | 12 | 1799 | |||
AK296551.1 | - | 12 | 1564 | |||
BC026320.1 | - | 13 | 1745 | 4797814 | ||
BP209348.1 | - | 1 | 52 |