Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018270.5 | NP_060740.1 | 3, 4 | 479 | ||
GenBank mRNA | AB085682.1 | - | 3, 4 | 379 | ||
AK001776.1 | - | 3, 4 | 431 | |||
BC009889.2 | - | 3, 4 | 398 | 4026378 | ||
BC014235.2 | - | 3, 4 | 403 | 4555060 | ||
BC018841.2 | - | 3, 4 | 398 | 3050866 | ||
BU902487.1 | - | 3, 4 | 462 |