Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303404.1 | NP_001290333.1 | 13 | 1626 | ||
NM_001303405.1 | NP_001290334.1 | 14 | 1791 | |||
NM_001303406.1 | NP_001290335.1 | 11 | 1480 | |||
NM_001303407.1 | NP_001290336.1 | 13 | 1515 | |||
NM_018294.5 | NP_060764.3 | 14 | 1746 | |||
GenBank mRNA | AK000876.1 | - | 4 | 586 | ||
AK001860.1 | - | 14 | 1692 | |||
AK023984.1 | - | 12 | 1343 | |||
AK097895.1 | - | 13 | 1514 | |||
AK225590.1 | - | 13 | 1584 | |||
AK295303.1 | - | 11 | 1450 | |||
AL832515.1 | - | 14 | 1703 | |||
BC008746.2 | - | 14 | 1687 | 3345869 | ||
BM476132.1 | - | 1 | 120 |