Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145029.1 | NP_001138501.1 | 36 | 4235 | ||
XM_006722325.3 | XP_006722388.1 | 42 | 4420 | |||
XM_011525662.2 | XP_011523964.1 | 42 | 4420 | |||
GenBank mRNA | AK294693.1 | - | 8 | 1627 |