Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 1894 | ||
NM_001145266.1 | NP_001138738.1 | 3 | 1797 | |||
NM_018389.4 | NP_060859.4 | 2 | 2168 | |||
XM_011520202.2 | XP_011518504.1 | 2 | 1055 | |||
GenBank mRNA | AF326199.1 | - | 2 | 2065 | ||
AK000527.1 | - | 1 | 713 | |||
AK074929.1 | - | 2 | 2169 | |||
BC001427.2 | - | 2 | 1473 | 3139599 |