Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025248.2 | NP_079524.2 | 19 | 6626 | ||
XM_017025169.1 | XP_016880658.1 | 23 | 7718 | |||
XM_017025170.1 | XP_016880659.1 | 21 | 6916 | |||
XM_017025171.1 | XP_016880660.1 | 20 | 6793 | |||
XM_017025172.1 | XP_016880661.1 | 19 | 6691 | |||
GenBank mRNA | BC019717.1 | - | 1 | 364 |