Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511671.2 | XP_011509973.1 | 6 | 1201 | ||
XM_011511672.2 | XP_011509974.1 | 5 | 1141 | |||
XM_011511673.2 | XP_011509975.1 | 6 | 1201 | |||
XM_017004736.1 | XP_016860225.1 | 5 | 1144 | |||
GenBank mRNA | BC041460.1 | - | 4 | 1291 |