Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015448.1 | 3 | 1811 | |||
GenBank mRNA | AK091367.1 | - | 2 | 1215 | ||
AK095619.1 | - | 3 | 1811 | |||
AK095764.1 | - | 1 | 14 | |||
BE263812.1 | - | 2 | 519 |