Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018930.3 | NP_061753.1 | 1 | 584 | ||
GenBank mRNA | AF152489.1 | - | 1 | 410 | ||
AF217748.1 | - | 1 | 410 | |||
AK056391.1 | - | 1 | 594 | |||
AK315583.1 | - | 1 | 610 | |||
AY358720.1 | - | 1 | 588 | |||
BC031837.1 | - | 1 | 584 | 4931995 |