Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042522.2 | NP_001035987.1 | 2 | 312 | ||
NR_073032.1 | 2 | 127 | ||||
XM_006723219.3 | XP_006723282.1 | 3 | 303 | |||
XM_011526978.2 | XP_011525280.1 | 2 | 358 | |||
GenBank mRNA | BC151136.1 | - | 2 | 312 | 9057036 | |
BF983829.1 | - | 2 | 129 | |||
DQ323927.1 | - | 2 | 213 | |||
DQ323928.1 | - | 2 | 213 |