Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC14A2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110899.1 | 4, 5 | 353 | |||
GenBank mRNA | AK126075.1 | - | 4, 5 | 353 |