Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018924.4 | NP_061747.2 | 1 | 2376 | ||
NM_032097.2 | NP_115268.2 | 1 | 2376 | |||
GenBank mRNA | AF152332.1 | - | 1 | 2245 | ||
AF152519.1 | - | 1 | 2245 | |||
BC150172.1 | - | 1 | 2376 | 40147143 |