Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 3 | 353 | ||
NM_019055.5 | NP_061928.4 | 3 | 559 | |||
XM_006718861.1 | XP_006718924.1 | 3 | 492 | |||
GenBank mRNA | AF361473.1 | - | 3 | 488 | ||
AK027852.1 | - | 3 | 458 | |||
AK074163.1 | - | 2 | 89 | |||
AK289769.1 | - | 3 | 458 | |||
AK302628.1 | - | 3 | 223 | |||
AY358083.1 | - | 3 | 410 | |||
BC014995.1 | - | 2 | 171 | |||
BC039602.1 | - | 3 | 447 | 5590503 | ||
BC111562.1 | - | 3 | 453 | 40068748 | ||
BC111748.1 | - | 3 | 453 | 40068752 | ||
DC424813.1 | - | 3 | 560 |