Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207020.1 | NP_001193949.1 | 4 | 1171 | ||
XM_006724254.3 | XP_006724317.1 | 4 | 1882 | |||
XM_006724255.2 | XP_006724318.1 | 4 | 1774 | |||
XM_006724256.3 | XP_006724319.1 | 4 | 1419 |