Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025190.3 | NP_079466.3 | 38 | 2961 | ||
XM_005263995.4 | XP_005264052.1 | 38 | 3241 | |||
XM_006712661.1 | XP_006712724.1 | 38 | 3241 | |||
XM_017004578.1 | XP_016860067.1 | 38 | 3238 | |||
XM_017004579.1 | XP_016860068.1 | 36 | 3139 | |||
XM_017004580.1 | XP_016860069.1 | 36 | 3139 | |||
XM_017004581.1 | XP_016860070.1 | 36 | 3139 | |||
XM_017004582.1 | XP_016860071.1 | 36 | 3139 | |||
XM_017004583.1 | XP_016860072.1 | 37 | 3103 | |||
XM_017004584.1 | XP_016860073.1 | 32 | 2935 | |||
XM_017004585.1 | XP_016860074.1 | 36 | 2996 | |||
XM_017004586.1 | XP_016860075.1 | 33 | 1982 | |||
XM_017004587.1 | XP_016860076.1 | 37 | 3170 | |||
XM_017004588.1 | XP_016860077.1 | 36 | 3035 | |||
XM_017004589.1 | XP_016860078.1 | 35 | 2928 | |||
XM_017004590.1 | XP_016860079.1 | 34 | 2855 | |||
XM_017004591.1 | XP_016860080.1 | 20 | 2959 | |||
XM_017004592.1 | XP_016860081.1 | 39 | 3285 | |||
XM_017004593.1 | XP_016860082.1 | 37 | 3091 | |||
XM_017004594.1 | XP_016860083.1 | 36 | 2984 | |||
XM_017004595.1 | XP_016860084.1 | 37 | 3040 | |||
GenBank mRNA | CA394381.1 | - | 3 | 337 |
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 83 | 5666 | ||
GenBank mRNA | AK123626.1 | - | 30 | 2514 |