Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098833.1 | NP_001092303.1 | 12 | 1391 | ||
NM_020218.1 | NP_064603.1 | 12 | 1412 | |||
XM_011525038.2 | XP_011523340.1 | 12 | 2063 | |||
XM_011525044.2 | XP_011523346.1 | 12 | 2006 | |||
XM_011525046.2 | XP_011523348.1 | 12 | 1985 | |||
XM_017024880.1 | XP_016880369.1 | 13 | 1641 | |||
XM_017024881.1 | XP_016880370.1 | 13 | 1940 | |||
XM_017024882.1 | XP_016880371.1 | 12 | 2042 | |||
XM_017024883.1 | XP_016880372.1 | 13 | 1644 | |||
XM_017024884.1 | XP_016880373.1 | 12 | 1666 | |||
XM_017024885.1 | XP_016880374.1 | 13 | 1587 | |||
XM_017024886.1 | XP_016880375.1 | 13 | 1566 | |||
GenBank mRNA | AK056002.1 | - | 9 | 935 | ||
AL390158.1 | - | 7 | 821 | |||
BC024695.2 | - | 5 | 749 | |||
BC037418.1 | - | 12 | 1319 |