Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281727.1 | NP_001268656.1 | 4 | 471 | ||
NM_001281728.1 | NP_001268657.1 | 5 | 550 | |||
NM_001281729.1 | NP_001268658.1 | 5 | 858 | |||
NM_014183.3 | NP_054902.1 | 4 | 475 | |||
NM_177953.2 | NP_808852.1 | 5 | 554 | |||
NR_104032.1 | 5 | 557 | ||||
GenBank mRNA | AF161511.1 | - | 4 | 446 | ||
AF165516.1 | - | 4 | 428 | |||
AK026864.1 | - | 3 | 411 | |||
BC002481.1 | - | 4 | 453 | 3347555 | ||
BG110161.1 | - | 4 | 460 | |||
BG419592.1 | - | 5 | 526 | |||
BM791453.1 | - | 5 | 536 | |||
BQ639804.1 | - | 5 | 523 | |||
BU163605.1 | - | 494 | ||||
CB149456.1 | - | 4 | 471 |