Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014238.1 | NP_055053.1 | 22 | 4203 | ||
XM_006722154.3 | XP_006722217.1 | 19 | 2862 | |||
XM_011525429.2 | XP_011523731.1 | 21 | 2970 | |||
XM_011525430.2 | XP_011523732.1 | 21 | 4357 | |||
XM_017025267.1 | XP_016880756.1 | 20 | 2928 | |||
XM_017025268.1 | XP_016880757.1 | 20 | 2904 | |||
XM_017025270.1 | XP_016880759.1 | 20 | 2705 | |||
XM_017025271.1 | XP_016880760.1 | 22 | 3677 | |||
XM_017025272.1 | XP_016880761.1 | 22 | 3660 | |||
XM_017025273.1 | XP_016880762.1 | 21 | 3483 | |||
XM_017025274.1 | XP_016880763.1 | 21 | 2686 | |||
XM_017025275.1 | XP_016880764.1 | 21 | 2673 | |||
XM_017025276.1 | XP_016880765.1 | 22 | 3625 | |||
XM_017025277.1 | XP_016880766.1 | 17 | 2368 | |||
XM_017025278.1 | XP_016880767.1 | 17 | 2378 |