Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181704.1 | NP_859055.1 | 357 | |||
GenBank mRNA | AF339515.1 | - | 357 |