Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 5 | 404 | ||
NM_001167741.1 | NP_001161213.1 | 4 | 334 | |||
XM_005248495.3 | XP_005248552.1 | 5 | 406 | |||
XM_017009440.1 | XP_016864929.1 | 4 | 328 | |||
GenBank mRNA | AK296864.1 | - | 5 | 399 | ||
BC150295.1 | - | 5 | 394 | |||
BX538064.1 | - | 4 | 314 | |||
BX647596.1 | - | 5 | 401 | |||
DB456588.1 | - | 5 | 404 |