siRNA Details
Gene Symbol: AGPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020133.2 | NP_064518.1 | 3 | 496 | ||
XM_006715514.3 | XP_006715577.1 | 3 | 482 | |||
XM_017011059.1 | XP_016866548.1 | 3 | 529 | |||
GenBank mRNA | AF156776.1 | - | 3 | 426 | ||
AJ227890.1 | - | 2 | 292 | |||
AJ227891.1 | - | 2 | 296 | |||
AJ420473.1 | - | 3 | 468 | |||
AK074208.1 | - | 3 | 482 | |||
AK295025.1 | - | 3 | 443 | |||
AK295548.1 | - | 3 | 440 | |||
AK297636.1 | - | 2 | 218 | |||
AY358506.1 | - | 3 | 436 | |||
BC020209.1 | - | 3 | 428 | 4850399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
