Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034944.1 | NP_001030116.1 | 2 | 177 | ||
GenBank mRNA | BC105895.1 | - | 2 | 177 | 7380480 |