Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031824.1 | NP_001026994.1 | 2 | 1212 | ||
GenBank mRNA | AB158417.1 | - | 2 | 1212 | ||
AB158418.1 | - | 2 | 1212 |