Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207633.2 | NP_997516.1 | 7 | 1404 | ||
GenBank mRNA | AK030473.1 | - | 7 | 1406 | ||
AK045453.1 | - | 7 | 1116 | |||
AK053562.1 | - | 7 | 1404 | |||
AK143152.1 | - | 7 | 974 | |||
BC048712.1 | - | 7 | 1039 | 6328657 |