Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020199.2 | NP_064584.1 | 3 | 1359 | ||
GenBank mRNA | AF035313.1 | - | 1 | 451 | ||
AF226055.1 | - | 3 | 1435 | |||
AK074963.1 | - | 3 | 1271 | |||
AK298515.1 | - | 4 | 1168 | |||
BC020875.1 | - | 3 | 1250 | 4610607 |