Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027357.2 | NP_081633.1 | 17, 18 | 2169 | ||
GenBank mRNA | AK010596.1 | - | 1, 2 | 116 | ||
AK087947.1 | - | 17, 18 | 2120 | |||
AK153386.1 | - | 17, 18 | 2170 | |||
AK159123.1 | - | 17, 18 | 2169 | |||
AK159750.1 | - | 17, 18 | 2148 | |||
BC018172.1 | - | 3, 4 | 218 | |||
BC022955.1 | - | 17, 18 | 2059 | |||
BC025802.1 | - | 17, 18 | 2073 | |||
BC029319.1 | - | 17, 18 | 2106 | |||
BC031587.1 | - | 17, 18 | 2100 | |||
BC032068.1 | - | 17, 18 | 2100 | |||
BC039937.1 | - | 17, 18 | 2146 | |||
BC089028.1 | - | 17, 18 | 2116 | |||
BC138526.1 | - | 17, 18 | 2132 | 8861546 | ||
BC138527.1 | - | 17, 18 | 2132 | 8861547 |