Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033194.2 | NP_001028366.1 | 18 | 2687 | ||
GenBank mRNA | AK145040.1 | - | 2691 | |||
AK160997.1 | - | 18 | 2676 | |||
AK165830.1 | - | 18 | 2688 | |||
BC141157.1 | - | 18 | 2677 | 9055769 |