Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020387.3 | NP_065120.2 | 5 | 1026 | ||
NR_133653.1 | 4 | 830 | ||||
GenBank mRNA | AF083124.1 | - | 5 | 984 | ||
AF274025.1 | - | 5 | 1005 | |||
BC004416.1 | - | 3 | 516 | |||
BC009831.2 | - | 5 | 1000 | 3926839 | ||
BC033322.1 | - | 5 | 990 | 4775725 |