Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC10A7 SIRNA Assays ›
Gene Symbol:
SLC10A7
Gene Name:
solute carrier family 10 member 7
Gene Aliases:
C4orf13, P7
Chromosome Location:
Chr.4: 146253981 - 146522351 on Build GRCh38
UniGene ID:
Hs.659209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
284842
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC10A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001029998.5
NP_001025169.1
1, 2
365
NM_001300842.2
NP_001287771.1
1, 2
365
NM_001317816.1
NP_001304745.1
1, 2
365
NM_001317817.1
NP_001304746.1
1, 2
365
NM_001317818.1
NP_001304747.1
1, 2
365
NM_032128.4
NP_115504.1
1, 2
365
NR_133924.1
1, 2
365
XM_011532311.2
XP_011530613.1
1, 2
365
XM_011532313.2
XP_011530615.1
11
XM_017008689.1
XP_016864178.1
1, 2
365
XM_017008690.1
XP_016864179.1
1, 2
365
XM_017008691.1
XP_016864180.1
1, 2
365
XM_017008692.1
XP_016864181.1
1, 2
745
GenBank mRNA
AK291046.1
-
1, 2
233
AL136728.1
-
1, 2
293
AY346324.1
-
1, 2
328
BC023288.1
-
1, 2
316
BC063471.1
-
1, 2
278
BC120970.1
-
1, 2
224
BC120971.1
-
1, 2
224
BC127626.1
-
1, 2
224
BC127627.1
-
1, 2
224
BC148252.1
-
1, 2
224
BC150308.1
-
1, 2
334
8860437
CR933647.1
-
1, 2
359
CR936751.1
-
1, 2
222
DA590625.1
-
1, 2
366
DB205503.1
-
1, 2
365
DQ122860.1
-
1, 2
111
DQ122861.1
-
1, 2
111
DQ871036.1
-
1, 2
111
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
transmembrane transport
symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0