Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKDD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182703.5 | NP_874362.3 | 15 | 2727 | ||
GenBank mRNA | BC042002.1 | - | 1 | 1102 | ||
BC047607.2 | - | 5 | 1734 | |||
BX116779.1 | - | 1 | 201 |