Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000111.1 | NP_001000111.1 | 1 | 336 |
Set Membership: |
G-protein coupled receptor |