Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020675.3 | NP_065726.1 | 3 | 295 | ||
XM_011511516.2 | XP_011509818.1 | 3 | 304 | |||
GenBank mRNA | AF225416.1 | - | 3 | 277 | ||
AK291093.1 | - | 3 | 306 | |||
AY456388.1 | - | 2 | 164 | |||
BC022255.1 | - | 3 | 295 | 4738187 | ||
CR457197.1 | - | 2 | 164 |