Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007777.3 | NP_031803.3 | 3 | 594 | ||
GenBank mRNA | AK014301.1 | - | 3 | 581 | ||
BC057012.1 | - | 565 | ||||
BC078417.1 | - | 569 | 6824205 | |||
BC132506.1 | - | 3 | 594 | 40130783 | ||
BC132508.1 | - | 3 | 594 | 40130785 | ||
K02584.1 | - | 548 |