Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291941.1 | NP_001278870.1 | 6 | 782 | ||
NM_001291942.1 | NP_001278871.1 | 5 | 670 | |||
NM_020685.4 | NP_065736.1 | 6 | 889 | |||
XM_017006931.1 | XP_016862420.1 | 6 | 1265 | |||
GenBank mRNA | AF236158.1 | - | 6 | 719 | ||
BC017772.2 | - | 5 | 699 | 4307022 |