Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290765.1 | NP_001277694.1 | 6 | 870 | ||
NM_026721.3 | NP_080997.1 | 6 | 831 | |||
GenBank mRNA | AK003191.1 | - | 2 | 133 | ||
AK035296.1 | - | 6 | 738 | |||
AK156951.1 | - | 6 | 756 | |||
BC020106.1 | - | 4 | 349 | |||
BC100463.1 | - | 6 | 838 | 30918840 | ||
BC125525.1 | - | 6 | 692 | 40130040 | ||
BC137752.1 | - | 6 | 692 | 8860771 |