Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tamm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026894.1 | NP_081170.1 | 5 | 740 | ||
GenBank mRNA | AK005100.1 | - | 5 | 740 | ||
AK165459.1 | - | 1 | 235 |