Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011543.2 | NP_001011543.2 | 5 | 546 | ||
NM_001251828.1 | NP_001238757.1 | 5 | 474 | |||
NM_021048.4 | NP_066386.2 | 4 | 455 | |||
GenBank mRNA | AK314161.1 | - | 4 | 455 | ||
BC004105.2 | - | 4 | 430 | 3926146 | ||
CB158974.1 | - | 5 | 514 | |||
CB959556.1 | - | 5 | 557 | |||
DA827990.1 | - | 5 | 436 |