Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000032.4 | NP_000023.2 | 5 | 577 | ||
NM_001037967.3 | NP_001033056.1 | 4 | 466 | |||
NM_001037968.3 | NP_001033057.1 | 5 | 573 | |||
XM_005261995.3 | XP_005262052.1 | 6 | 597 | |||
XM_017029354.1 | XP_016884843.1 | 6 | 598 | |||
GenBank mRNA | AK290565.1 | - | 4 | 450 | ||
AK291589.1 | - | 4 | 379 | |||
AK313118.1 | - | 5 | 577 | |||
BC030230.2 | - | 4 | 452 | 5226194 | ||
BP284609.1 | - | 4 | 466 | |||
X56352.1 | - | 4 | 436 | |||
X60364.1 | - | 5 | 491 |