Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRB10 SIRNA Assays ›
Gene Symbol:
GRB10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location:
Chr.7: 50590063 - 50793462 on Build GRCh38
UniGene ID:
Hs.164060
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289529
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRB10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001549.2
NP_001001549.1
4
458
NM_001001550.2
NP_001001550.1
4
538
NM_001001555.2
NP_001001555.1
5
785
NM_005311.4
NP_005302.3
4
458
XM_011515304.1
XP_011513606.1
7
897
XM_011515312.2
XP_011513614.1
6
1185
XM_011515316.1
XP_011513618.1
5
730
XM_017012029.1
XP_016867518.1
5
982
XM_017012030.1
XP_016867519.1
4
916
XM_017012031.1
XP_016867520.1
5
982
XM_017012032.1
XP_016867521.1
10
1359
XM_017012033.1
XP_016867522.1
7
897
XM_017012034.1
XP_016867523.1
5
498
XM_017012035.1
XP_016867524.1
11
2408
XM_017012036.1
XP_016867525.1
12
2193
XM_017012037.1
XP_016867526.1
12
2030
XM_017012038.1
XP_016867527.1
11
1980
XM_017012039.1
XP_016867528.1
8
1227
XM_017012040.1
XP_016867529.1
8
1763
XM_017012041.1
XP_016867530.1
10
1884
XM_017012042.1
XP_016867531.1
6
804
XM_017012043.1
XP_016867532.1
7
994
XM_017012044.1
XP_016867533.1
4
728
XM_017012045.1
XP_016867534.1
4
392
XM_017012046.1
XP_016867535.1
8
1744
XM_017012047.1
XP_016867536.1
12
2592
XM_017012048.1
XP_016867537.1
11
2476
XM_017012049.1
XP_016867538.1
12
2098
XM_017012050.1
XP_016867539.1
11
2048
XM_017012051.1
XP_016867540.1
10
1952
XM_017012052.1
XP_016867541.1
10
2292
XM_017012053.1
XP_016867542.1
11
1914
XM_017012054.1
XP_016867543.1
10
1864
XM_017012055.1
XP_016867544.1
7
1647
XM_017012056.1
XP_016867545.1
6
2479
XM_017012057.1
XP_016867546.1
9
1768
XM_017012058.1
XP_016867547.1
5
639
XM_017012059.1
XP_016867548.1
5
761
XM_017012060.1
XP_016867549.1
13
2725
XM_017012061.1
XP_016867550.1
4
591
XM_017012062.1
XP_016867551.1
3
352
XM_017012063.1
XP_016867552.1
12
2592
XM_017012064.1
XP_016867553.1
11
2476
XM_017012065.1
XP_016867554.1
9
1928
XM_017012066.1
XP_016867555.1
7
2287
XM_017012067.1
XP_016867556.1
7
790
XM_017012068.1
XP_016867557.1
7
1656
GenBank mRNA
AB000731.1
-
4
433
AF000017.1
-
4
427
AF001534.1
-
4
458
AJ271366.1
-
7
897
AK289511.1
-
6
752
BC024285.1
-
5
562
4590330
BX649028.1
-
4
518
D86962.1
-
6
1190
U34355.1
-
4
458
U66065.1
-
4
538
U69276.1
-
4
540
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of phosphorylation
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0