Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005332.2 | NP_005323.1 | 2 | 270 | ||
XM_005255287.3 | XP_005255344.1 | 3 | 900 | |||
XM_005255288.3 | XP_005255345.1 | 3 | 1017 | |||
GenBank mRNA | BC027892.1 | - | 2 | 250 | 5224569 | |
CR456848.1 | - | 2 | 215 | |||
M24173.1 | - | 2 | 244 |